Inch Pound Fixed Head Torque Wrenches, Proto®, ORS Nasco
Supplier: Safety & Industrial Supplies
577-6063C 577-6065C
300027-280EA 392.12 USD
300027-280 300027-284
Inch Pound Fixed Head Torque Wrenches, Proto®, ORS Nasco
Wrenches
Manufactured to meet standard industrial torque applications, general industry, heavy equipment, and maintenance.
  • Calibrated to +/- 3% clockwise direction and +/- 6% counter clockwise at 20% to 100% of full scale
  • Calibration is accomplished by manual loading the torque wrenches