Click-Stop® Adjustable Wrenches, Proto®
Supplier: Safety & Industrial Supplies
577-715L 577-706L 577-712L
300003-082EA 132.92 USD
300003-082 300027-528 300027-540
Click-Stop® Adjustable Wrenches, Proto®
Wrenches
Clik-Stop® automatic locking jaw, locks jaw at any opening you select.
  • Extra wide jaw capacity
  • Large serrated knurl for jaw adjustment
  • Thin, slightly tapered jaw for access in tight work areas
  • Square jaw opening for greater strength
  • Radius corner design increases strength and fastener surface contact area