Spatulas, Ampco Safety Tools
Supplier: Safety & Industrial Supplies
065-S-91 065-S-92 065-S-90
300012-718EA 78.67 USD
300012-718 300012-720 300012-716
Spatulas, Ampco Safety Tools
Spatulas
These spatulas are made of high strength nickel tin copper blade material.
  • Non-sparking
  • Non-magnetic
  • Corrosion resistant
  • Beryllium-free