Standard Cable Ties, GARDNER BENDER, ORS Nasco
Supplier: Safety & Industrial Supplies
623-46-310M 623-46-315 623-46-310 623-46-315M 623-46-308 623-46-308MN 623-46-308UVB
300090-676PK 130.38 USD
300090-676 300090-680 300090-674 300090-682 300090-668 300090-670 300090-672
Standard Cable Ties, GARDNER BENDER, ORS Nasco
Ties
Cable ties with doubleLock design has teeth on both sides, ensuring 50% greater strength than industry standards.
  • Made from durable 6/6 nylon material
  • Serviceable through temperatures of 32 to 185 °F
  • UVB ties resist harmful light rays
  • 20.3 cm (8") ties close to a maximum diameter of 5.1 cm (2")
  • 27.9 cm (11") ties close to a maximum diameter of 7.6 cm (3")
  • 35.6 cm (14") ties close to a maximum diameter of 10.2 cm (4")
It used for bundling wire, cable, hose, ducts, and countless other applications.