Original Formula Cleaners, Simple Green®
Supplier: Simple Green
Simple Green®
676-13012 676-13005
300050-364CS 282.41 USD
300050-364 300050-354
Original Formula Cleaners, Simple Green®
Cleaning Agents Detergents and Surface Cleansers
Butyl cellosolve cleaners offer a safer alternative to toxic cleaners, solvents, and bleaches. Can be used on any washable, indoor, outdoor, or automotive surface.
  • Safer alternative to toxic cleaners, solvents, and bleaches
  • Can be used on any washable surface
Acidity: 9.35 pH