PharmaSafety™ Sharps Disposal Containers, Covidien
Supplier: Covidien
PharmaSafety™
8860- 8850 8836SA 8871 8870- 8820
95043-682CS 322.13 USD
95043-682 95043-680 95043-692 95043-690 95043-688 95043-686
PharmaSafety™ Sharps Disposal Containers, Covidien
Bins Sharps Containers
Developed to meet state regulations for pharmaceutical waste, containers feature leak-resistant gaskets and absorbent pads to contain liquid contents
They are white with blue lids to distinguish them from other disposal containers. Supplied with tamper-evident labels.