Scotch® Vinyl Electrical Color Coding Tapes 35, ORS Nasco, INC.
Supplier: ORS Nasco
Scotch®
500-10836 500-10810 7000058435 500-11271 500-10828 500-10869 500-10851
300009-331EA 17.16 USD
300009-331 300009-332 300009-335 300009-336 300009-333 76242-414 300009-338
Scotch® Vinyl Electrical Color Coding Tapes 35, ORS Nasco, INC.
Tapes
Designed for use in phase identification, color coding of motor leads and piping systems, and for marking safety areas.
  • Excellent electrical insulating properties
  • Wraps smoothly, conforms well, and holds securely over a wide range of temperatures
  • Polyvinyl chloride (PVC) backing
  • Pressure sensitive rubber based adhesive
  • Inhibits corrosion of electrical conductors