Scotch® Super Vinyl Electrical Tapes 88, ORS Nasco, INC.
Supplier: ORS Nasco
Scotch®
500-10356 500-10364 500-06143
300009-330EA 16.6 USD
300009-330 300009-322 300009-329
Scotch® Super Vinyl Electrical Tapes 88, ORS Nasco, INC.
Tapes
Scotch® Super vinyl electrical tapes are compatible with solid dielectric cable insulations, rubber and synthetic splicing compounds, and epoxy and polyurethane resins.
  • Polyvinyl chloride (PVC) backing
  • Pressure sensitive rubber based adhesive
  • Inhibits corrosion of electrical conductors