Scotch® Super Vinyl Electrical Tapes 33+, ORS Nasco, INC.
Supplier: ORS Nasco
Scotch®
500-06133 500-10075 500-06130
300009-321EA 16.44 USD
300009-321 300009-327 300009-319
Scotch® Super Vinyl Electrical Tapes 33+, ORS Nasco, INC.
Tapes
Scotch® Super Vinyl tape provides moisture-tight electrical and mechanical protection with minimum bulk.
  • Resists abrasion, corrosion, alkalies, and ultraviolet light
  • Withstands temperature up to 104 °C (220 °F)
Pressure-sensitive rubber based adhesive is compatible with solid dielectric cable insulation, rubber and synthetic splicing compounds, and epoxy or polyurethane resins.