Phosphate Buffer Solutions for Hematology, Azer Scientific
Supplier: AZER SCIENTIFIC INC
Low molarity phosphate buffers optimized for use with Azer Scientific Wright and Wright-Giemsa stain solutions.
- Suitable for use with any Wright, Wright-Giemsa, or Giemsa stain solutions
- Low molar strength formula extends the life of stain dilutions
- Available in pH range from 6.6-7.0 to achieve desired stain intensity and colors
Ready to use phosphate buffer optimized for use in manual and automated hematology stain protocols.