Thermo Scientific™ MAS™ chemTRAK™ H General Chemistry Controls, Thermo Scientific
Supplier: MICROGENICS CORPORATION (MAS)
CHA-1 CHA-2 CHU-1 CHU-2 CHA-3 CHU-3 10014151 10014152 10014150
89130-610EA 291.87 USD
89130-610 89130-612 89130-616 89130-618 89130-614 89130-620 10154-160 10154-162 10154-290
Thermo Scientific™ MAS™ chemTRAK™ H General Chemistry Controls, Thermo Scientific
Controls and Calibration Verification Materials
Ensure accurate results in general chemistry and TDM methods with comprehensive, multi-analyte MAS™ chemTRAK™ H General Chemistry Controls
Human serum matrix. Assayed values available for most major analyzers. Refer to package insert for specific analyte and stability claims.
Analytes: Acetaminophen, ACP, ALT, Albumin, ALP, Amikacin, Amylase, Apolipoprotein A-1, Apolipoprotein B, AST, CO2, Bilirubin, Direct, Bilirubin, Total, BUN, C3 Complement, C4 Complement, Caffeine, Calcium, Carbamazepine, Chloride, Cholesterol, Cholesterol HDL, Cholesterol LDL, Cholinesterase, Creatine Kinase (CK), Creatinine, Lipoprotein, Digoxin, Disopyramide, Ethanol, Ethosuximide, Ferritin, GGT, Gentamicin, Glucose, GLDH, Haptoglobin, HBDH, IgA, IgG, IgM, Iron, TIBC, UIBC, LDH, Lactic Acid, Lidocaine, Lipase, Lithium, Magnesium, Methotrexate, NAPA, Osmolality, Phenobarbital, Phenytoin, Phosphorus, Potassium, Prealbumin, Prmidone, Procainamide, Quinidine, Salicylate, Sodium, T3, T4, Theophylline, hTSH, Tobramycin, Total Protein, Transferrin, Tricyclic Antidepressants, Triglycerides, T-Uptake, Uric Acid, Valproic Acid, Vancomycin. (refer to package insert for specific analyte and stability claims).
These liquid, ready-to-use controls are available in a convenient Vista™ vial format in both assayed and unassayed formats.
Ordering information: Available in Vista™ vial format, Comprehensive, multi-analyte chemistry/TDM control.