MAS™ Bilirubin Controls, Thermo Scientific
Supplier: MICROGENICS CORPORATION (MAS)
Monitor the performance of total, conjugated, unconjugated and neonatal Bilirubin with tri-level, assayed MAS™ Bilirubin Controls
Liquid, ready-to-use; no additional preparation steps required; Bovine serum matrix; Assayed values available for most major analyzers; Elevated bilirubin in Level 3 for neonatal applications.
Analytes: Conjugate Bilirubin, Total Bilirubin, Direct Bilirubin, Unconjugated Bilirubin, Neonatal Bilirubin.
These controls are also available in a convenient Vista™ vial format.
Ordering information: Available in individual levels only.