BD™ Whitacre Pencil Point Spinal Needles, BD Medical
Supplier: BD Medical
BD™
405079 405010
BD405079CS 1038.47 USD
BD405079 BD405010
BD™ Whitacre Pencil Point Spinal Needles, BD Medical
Syringe Needles
Sterile, single-use needles feature a pencil point configuration that spreads dural fibers to minimize CSF loss and reduce the occurrence of post-procedure headaches
The needles advance straight through ligaments for on-target access and drug delivery, and make a distinct “pop” as the point penetrates the dura. High-strength stainless steel with special etching assures a smooth, sensitive feel and a low-glare hub provides fast, accurate visualization of CSF.