Seracult® Fecal Occult Blood Tests, Propper Manufacturing
Supplier: Propper
Seracult®
37901500 37200500 37901600 37100100 37901000 37100200 37200300 37901700 37200400
10753-456CS 660.1 USD
10753-456 14230-050 60829-035 10753-472 14230-049 14230-027 10753-470 14230-039 14230-048
Seracult® Fecal Occult Blood Tests, Propper Manufacturing
Clinical Diagnostic Supplies Clinical Reagents and Kits
Seracult® fecal occult blood test is a qualitative aid in the diagnosis of colorectal cancer.
  • Guaiac fecal occult blood test to aid in the detection of colorectal cancer
  • Single or Triple Slide
  • Long Sample Stability
  • Fast qualitative results
  • Mailing kits of in-office slides
  • 28 days sample stability
Seracult® Fecal Occult Blood Test is a qualitative aid in the diagnosis of colorectal cancer. Designed for routine use in physical examinations in doctors offices, hospitals, clinics, and laboratories or in the privacy and convenience of a patient's home. Seracult® is available in a variety of configurations to meet all of your needs. Seracult paper allows for enhanced readability for early detection of even the smallest amount of blood in the sample.