Acetic anhydride 97+% ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Acetyl oxide
, Acetyl acetate
AC42323-5000
200004-064EA
77.42
USD
200004-064
Acetic anhydride 97+% ACS
Acetic anhydride
Formula:
(CH₃CO)₂O MW: 102.09 g/mol Boiling Pt: 139.9 °C (1013 hPa) Melting Pt: –73.1 °C Density: 1.087 g/cm³ Flash Pt: 54 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008705 CAS Number: 108-24-7 EINECS: 203-564-8 UN: 1715 ADR: 8,II REACH: 01-2119486470-36 Merck Index: 13,00057 |
Specification Test Results
Appearance | Clear liquid |
GC | ≥97.0 % |
Color scale | ≤10 APHA |
Residue after evaporation | ≤0.003 % |
Chloride (Cl) | ≤5 ppm |
Phosphate (PO4) | ≤10 ppm |
Sulfate (SO4) | ≤5 ppm |
Iron (Fe) | ≤5 ppm |
Heavy metals (as Pb) | ≤2 ppm |
KMnO4-reducing substance | Passes test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...