Dithiothreitol (DTT, Cleland's reagent) 98%, pure

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (R*,R*)-1,4-Dimercapto-2,3-butandiol DTT

AC40919-1000 AC40919-0010 AC40919-0050 AC40919-0250
102777-710EA 503.45 USD
102777-710 200026-688 200014-352 102777-918
Dithiothreitol (DTT, Cleland's reagent) 98%, pure
Dithiothreitol (DTT, Cleland's reagent)
Formula: C₄H₁₀O₂S₂
MW: 154.25 g/mol
Boiling Pt: 125…130 °C (2 mmHg)
Melting Pt: 39…44 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004877
CAS Number: 3483-12-3
EINECS: 222-468-7
UN: 3335
ADR: 9,III
https://us.vwr.com/stibo/web/std.lang.all/98/19/2299819.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR