Bismuth 99.5%, powder -100 mesh

Supplier: Thermo Scientific Chemicals
Danger

AC31809-1000
200011-426EA 52.23 USD
200011-426
Bismuth 99.5%, powder -100 mesh
Bismuth
Formula: Bi
MW: 208.98 g/mol
Melting Pt: 271 °C
Storage Temperature: Ambient
MDL Number: MFCD00134033
CAS Number: 7440-69-9
EINECS: 231-177-4
UN: 3089
ADR: 4.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR