Potassium sorbate 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Potassium (2E,4E)-hexa-2,4-dienoate
, Potassium 2,4-Hexadienoate
, Sorbic acid potassium salt
AC29174-0010
AC29174-2500
AC29174-0050
AC29174-1000
103517-778EA
429
USD
103517-778
200007-472
103517-782
103517-780
Potassium sorbate 99%
Potassium sorbate
Formula:
CH₃CH=CHCH=CHCOOK MW: 150.22 g/mol Melting Pt: 270 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00016546 CAS Number: 24634-61-5 EINECS: 246-376-1 Merck Index: 13,07756 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...