2-Nitrobenzoyl chloride 95%

Supplier: Thermo Scientific Chemicals
Danger

AC23231-0250 AC23231-0050 AC23231-1000
200005-134EA 116 USD
200005-134 200016-722 103531-936
2-Nitrobenzoyl chloride 95%
2-Nitrobenzoyl chloride
Formula: C₇H₄ClNO₃
MW: 185.57 g/mol
Boiling Pt: 148…149 °C (9 mmHg)
Melting Pt: 17…20 °C
Density: 1.4 g/cm³
Storage Temperature: Refrigerator
MDL Number: MFCD00007131
CAS Number: 610-14-0
EINECS: 210-206-4
UN: 1760
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear yellow to orange to brown Liquid after melting
Infrared spectrum Conforms
Refractive index 1.5665 to 1.5695 (20 °C, 589 nm)
HPLC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR