3-Methyl-2-buten-1-ol 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2-Methyl-2-buten-4-ol
AC17192-0050
AC17192-1000
AC17192-5000
102776-368EA
150
USD
102776-368
200005-270
102774-876
3-Methyl-2-buten-1-ol 99%
3-Methyl-2-buten-1-ol
Formula:
C₅H₁₀O MW: 86.13 g/mol Boiling Pt: 140 °C (1013 hPa) Melting Pt: –86 °C Density: 0.84 g/cm³ Flash Pt: 43 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002916 CAS Number: 556-82-1 EINECS: 209-141-4 UN: 1987 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...