Tributyl phosphate 99+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
TBP
, TBPA
AC16993-0100
AC16993-0025
AC16993-0010
200001-686EA
77.22
USD
200001-686
200007-934
103466-678
Tributyl phosphate 99+%
Tributyl phosphate
Formula:
(CH₃(CH₂)₃O)₃PO MW: 266.32 g/mol Boiling Pt: 289 °C (760 mmHg) Melting Pt: –79 °C Density: 0.979 g/cm³ Flash Pt: 146 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009436 CAS Number: 126-73-8 EINECS: 204-800-2 Merck Index: 12,09749 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...