L(+)-Glutamic acid 99%

Supplier: Thermo Scientific Chemicals

Synonyms: L-Glutamic acid(S)-(+)-1-Aminopropane-1,3-dicarboxylic acidL(+)-α-Glutamic acidH-Glu-OH(S)-(+)-Glutamic acid(S)-(+)-α-Glutamic acidL(+)-1-Aminopropane-1,3-dicarboxylic acid(S)-(+)-2-Aminopentanedioic acidL(+)-2-Aminopentanedioic acid

AC15621-1000 AC15621-0050 AC15621-2500 AC15621-0010
103510-652EA 295.34 USD
103510-652 200006-824 200006-822 200006-820
L(+)-Glutamic acid 99%
L(+)-Glutamic acid
Formula: HO₂CCH₂CH₂CH(NH₂)CO₂H
MW: 147.13 g/mol
Melting Pt: 205 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00002634
CAS Number: 56-86-0
EINECS: 200-293-7
Merck Index: 13,04482
https://us.vwr.com/stibo/web/std.lang.all/98/27/2299827.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR