1-Dodecanol 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Dodecyl Alcohol
, 1-Dodecyl alcohol
, 1-Hydroxy dodecane
, Undecyl carbinol
, Lauryl alcohol
AC15545-0100
AC15545-0010
AC15545-0050
AC15545-5000
200008-658EA
37.4
USD
200008-658
200004-724
200001-306
103525-856
1-Dodecanol 98%
1-Dodecanol
Formula:
H₃C(CH₂)₁₀CH₂OH MW: 186.34 g/mol Boiling Pt: 258…265 °C (760 mmHg) Melting Pt: 22…27 °C Density: 0.835 g/cm³ Flash Pt: 119 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004753 CAS Number: 112-53-8 EINECS: 203-982-0 UN: 3077 ADR: 9,III Merck Index: 13,03439 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...