EDTA trisodium salt hydrate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Ethylenedinitrilotetra-acetic acid trisodium salt hydrate
, Ethylenediaminetetra-acetic acid trisodium salt hydrate
, Trisodium hydrogen ethylenediaminetetraacetate hydrate
, Edetic acid trisodium salt hydrate
AC14786-5000
200012-484EA
99.94
USD
200012-484
EDTA trisodium salt hydrate 98%
EDTA trisodium salt hydrate
Formula:
C₁₀H₁₃N₂Na₃O₈·H₂O MW: 376.21 g/mol Melting Pt: 300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00149675 CAS Number: 85715-60-2 EINECS: 205-758-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...