L(-)-Tryptophan 99%

Supplier: Thermo Scientific Chemicals

Synonyms: L-α-Amino-3-indolepropionic acid(S)-2-Amino-3-(3-indolyl)propionic acidH-Trp-OHL-TryptophanTryptophan

AC14059-1000 AC14059-0250 AC14059-0050 AC14059-5000
200000-982EA 47.11 USD
200000-982 200006-766 200006-764 200031-662
L(-)-Tryptophan 99%
L(-)-Tryptophan
Formula: C₁₁H₁₂N₂O₂
MW: 204.23 g/mol
Melting Pt: 280…285 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00064340
CAS Number: 73-22-3
EINECS: 200-795-6
Merck Index: 12,09929

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR