2-Thiophenemethanol 99+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Thiophen-2-ylmethanolThiophen-2-methanol2-Thienylmethanol

AC13886-0250
200016-770EA 70.11 USD
200016-770
2-Thiophenemethanol 99+%
2-Thiophenemethanol
Formula: C₅H₆OS
MW: 114.17 g/mol
Boiling Pt: 206…207 °C (761 mmHg)
Density: 0.9 g/cm³
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005454
CAS Number: 636-72-6
EINECS: 211-264-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR