2-Methylquinoxaline 97%

Supplier: Thermo Scientific Chemicals
Warning

AC12766-1000 AC12766-0050 AC12766-0250
200041-186EA 92.08 USD
200041-186 200041-188 103464-862
2-Methylquinoxaline 97%
2-Methylquinoxaline
Formula: C₉H₈N₂
MW: 144.18 g/mol
Boiling Pt: 245…247 °C (760 mmHg)
Density: 1.11 g/cm³
Flash Pt: 107 °C
Storage Temperature: Ambient
MDL Number: MFCD00006727
CAS Number: 7251-61-8
EINECS: 230-664-9

Order Now

Specification Test Results

Appearance Clear orange to dark red Liquid (May darken on storage)
Infrared spectrum Conforms
GC ≥96.0 %
GC ≥95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR