1,6-Hexanedithiol 97%

Supplier: Thermo Scientific Chemicals

Synonyms: 1,6-Dimercaptohexane

AC12067-0250 AC12067-0050
103509-572EA 123.85 USD
103509-572 200015-628
1,6-Hexanedithiol 97%
1,6-Hexanedithiol
Formula: C₆H₁₄S₂
MW: 150.31 g/mol
Boiling Pt: 237 °C (760 mmHg)
Melting Pt: –21 °C
Density: 0.99 g/cm³
Flash Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00004910
CAS Number: 1191-43-1
EINECS: 214-735-1
UN: 3334
ADR: 9,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5110 to 1.5150 (20 °C, 589 nm)
GC ≥96.0 %
Specific optical rotation +16° to +18° (20 °C, 589 nm) (c=1,H2O)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR