N-Ethyldiisopropylamine ≥98%, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
N.N’-Diisopropyl ethylamine
, N,N-Diisopropylethylamine
, DIPEA
, Ethyldiisopropylamine
, Hünig’s base
M-Clarity™ quality level = MQ200
8.00894.9150
8.00894.9019
8.00894.9019
8.00894.0100
EM8.00894.9019EA
1329.27
USD
EM8.00894.9019
CA8.00894.9019
EM8.00894.9150
CA8.00894.0100
N-Ethyldiisopropylamine ≥98%, Sigma-Aldrich®
N-Ethyldiisopropylamine
Formula:
C₈H₁₉N MW: 129.25 g/mol Melting Pt: 127 °C (1013 hPa) Density: 0.76 g/cm3 at 20 °C Flash Pt: 6 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008868 CAS Number: 7087-68-5 EINECS: 230-392-0 UN: 3384 ADR: 6.1,3,I |
Specification Test Results
Assay (GC, area %) | 98% min |
Density (d 20°/4°) | 0.755-0.758 |
Identity (IR-spectrum) | Conforms |
Water (H2O) | 0.2% max |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...