1-(Difluoromethoxy)-3-nitrobenzene 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-(Difluoromethoxy)nitrobenzene

B22267-14 B22267-06 B22267-06 B22267-14
AAB22267-06EA 61.84 USD
AAB22267-06 CAAAB22267-14 CAAAB22267-06 AAB22267-14
1-(Difluoromethoxy)-3-nitrobenzene 98%
1-(Difluoromethoxy)-3-nitrobenzene
Formula: C₇H₅F₂NO₃
MW: 189.12 g/mol
Boiling Pt: 240…242 °C
Density: 1.391
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD03407974
CAS Number: 22236-07-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR