(±)-Camphene (sum of camphene + fenchene), tech. (sum of camphene + fenchene)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (±)-3,3-Dimethyl-2-methylennorcamphaneCamphene(±)-2,2-Dimethyl-3-methylenebicyclo[2.2.1]heptane

A18040-0B A18040-0B
AAA18040-0BEA 203 USD
CAAAA18040-0B AAA18040-0B
(±)-Camphene (sum of camphene + fenchene), tech. (sum of camphene + fenchene)
(±)-Camphene
Formula: C₁₀H₁₆
MW: 136.24 g/mol
Boiling Pt: 159…160 °C
Melting Pt: 42…45 °C
Density: 0.850
Flash Pt: 34 °C(94 °F)
Storage Temperature: Ambient
MDL Number: MFCD00066603
CAS Number: 79-92-5
UN: 1325
ADR: "4.1",III
Merck Index: 14,1730

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR