Squalane 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
2,6,10,15,19,23-Hexamethyltetracosane
A17931-36
A17931-22
AAA17931-36EA
228.65
USD
AAA17931-36
AAA17931-22
Squalane 98%
Squalane
Purity: 98%
CAS No.: 111-01-3
Molecular Formula: C30H62
Formula Weight: 422.83
Flash Point: 218°(424°F)
Refractive Index: 1.4510
Melting Point: -38°
Boiling Point: 210-215°/1mm
Beilstein Reference: 776019
Merck Reference: 14,8767
MDL No.: MFCD00008953
Formula:
C₃₀H₆₂ MW: 422.82 g/mol Boiling Pt: 470 Melting Pt: -35 Density: 0.81 Flash Pt: 218 °C (424 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008953 CAS Number: 111-01-3 EINECS: 203-825-6 Merck Index: 14,8767 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...