Guaifenesin 99+%

Supplier: Thermo Scientific Chemicals
Warning

A16827-30 A16827-18
AAA16827-30EA 107 USD
AAA16827-30 AAA16827-0B AAA16827-18
Guaifenesin 99+%
Guaifenesin

Centrally acting muscle relaxant with expectorant properties


Purity: 99+% CAS No.: 93-14-1 Molecular Formula: C10H14O4 Formula Weight: 198.22 Melting Point: 80-82° Boiling Point: 215°/19mm Merck Reference: 14,4555 MDL No.: MFCD00016873

Formula: C₁₀H₁₄O₄
Boiling Pt: 215 °C/19 mm
Melting Pt: 80…82 °C
Storage Temperature: Ambient
MDL Number: MFCD00016873
CAS Number: 93-14-1
EINECS: 202-222-5
Merck Index: 14,4555

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR