1-Bromotetradecane 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Myristyl bromide

A16082-22 A16082-36 A16082-22 A16082-0E A16082-0E A16082-36
AAA16082-0EEA 342 USD
CAAAA16082-36 CAAAA16082-22 AAA16082-0E AAA16082-36 CAAAA16082-0E AAA16082-22
1-Bromotetradecane 98%
1-Bromotetradecane
Formula: BrH₂C(CH₂)₁₂CH₃
MW: 277.29 g/mol
Boiling Pt: 175…178 °C/20 mm
Melting Pt: 4…6 °C
Density: 1.018
Flash Pt: 158 °C(316 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000228
CAS Number: 112-71-0
EINECS: 203-999-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR