2-Thiophenemethanol 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Thienylmethanol Thiophen-2-methanol

A15768-14 A15768-14 A15768-36 A15768-22 A15768-36 A15768-22
AAA15768-22EA 154.99 USD
CAAAA15768-14 CAAAA15768-36 AAA15768-22 AAA15768-14 AAA15768-36 CAAAA15768-22
2-Thiophenemethanol 97%
2-Thiophenemethanol
Formula: C₅H₆OS
MW: 114.17 g/mol
Boiling Pt: 206…207 °C
Melting Pt: 0 °C
Density: 1.206
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005454
CAS Number: 636-72-6
EINECS: 211-264-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR