Cyanuric acid 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,3,5-Triazinetrion, 1,3,5-Triazine-2,4,6-triol, 1,3,5-Triazinane-2,4,6-trione, Isocyanuric acid

A15447-0B A15447-22 A15447-30
AAA15447-0BEA 134.93 USD
AAA15447-0B AAAA15447-22 AAA15447-30
Cyanuric acid 99%
Cyanuric acid

Purity: 99% CAS No.: 108-80-5 Molecular Formula: C3H3N3O3 Formula Weight: 129.08 Melting Point: 360° Beilstein Reference: 126982 Merck Reference: 14,2698 MDL No.: MFCD00082990

Formula: C₃H₃N₃O₃
MW: 129.08 g/mol
Melting Pt: >360 °C
Density: 2.5
Storage Temperature: Ambient
MDL Number: MFCD00082990
CAS Number: 108-80-5
EINECS: 203-618-0
Merck Index: 14,2698

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR