4-Propylphenol 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: p-Propylphenol, 4-n-Propylphenol

A15342-22 A15342-06 A15342-14
AAAA15342-22EA 521 USD
AAAA15342-22 AAA15342-06 AAA15342-14
4-Propylphenol 98%
4-Propylphenol

Purity: 98% CAS No.: 645-56-7 Molecular Formula: C9H12O Formula Weight: 136.19 Flash Point: 106°(222°F) Refractive Index: 1.5230 Melting Point: 20-22° Boiling Point: 232° Beilstein Reference: 1363429 UN No.: 3145 MDL No.: MFCD00002395

Formula: C₉H₁₂O
MW: 136.19 g/mol
Boiling Pt: 232
Melting Pt: 24
Density: 0.983
Flash Pt: 106 °C (222 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002395
CAS Number: 645-56-7
EINECS: 211-446-2
UN: 3145
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR