Quinoxaline 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Used as dyes, pharmaceuticals and antibiotics intermediate
Purity: 98+%
CAS No.: 91-19-0
Molecular Formula: C8H6N2
Formula Weight: 130.15
Flash Point: 98°(208°F)
Melting Point: 27-32°
Boiling Point: 220-223°
Beilstein Reference: 109351
Merck Reference: 14,8078
MDL No.: MFCD00006719
Formula:
C₈H₆N₂ Boiling Pt: 220…223 °C Melting Pt: 27…32 °C Density: 1.124 Flash Pt: 98 °C(208 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006719 CAS Number: 91-19-0 EINECS: 202-047-4 Merck Index: 14,8078 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...