Pentafluorobenzoic acid 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12396-06
A12396-14
AAA12396-06EA
41.7
USD
AAA12396-06
AAA12396-14
AAA12396-22
Pentafluorobenzoic acid 99%
Pentafluorobenzoic acid
Purity: 99%
CAS No.: 602-94-8
Molecular Formula: C7HF5O2
Formula Weight: 212.07
Melting Point: 100-104°
Boiling Point: 219-221°
MDL No.: MFCD00002406
Formula:
C₇HF₅O₂ Boiling Pt: 219…221 °C Melting Pt: 100…104 °C Density: 1,942 g/cm³ (22 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002406 CAS Number: 602-94-8 EINECS: 210-026-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...