Dimethylammonium chloride 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Dimethylamine hydrochloride

A12133-0B A12133-0I A12133-30
AAA12133-0BEA 89.9 USD
AAA12133-0B AAA12133-0I AAA12133-30
Dimethylammonium chloride 98+%
Dimethylammonium chloride

MDL: MFCD00012477 Beilstein Registry No.: 3589311


Purity: 98+% CAS No.: 506-59-2 Molecular Formula: (CH3)2NH.HCl Formula Weight: 81.55 Melting Point: 168-174° Beilstein Reference: 3589311 Merck Reference: 14,3228 MDL No.: MFCD00012477

Formula: C₂H₈ClN
MW: 81.54 g/mol
Melting Pt: 168…174 °C
Storage Temperature: Ambient
MDL Number: MFCD00012477
CAS Number: 506-59-2
EINECS: 208-046-5
Merck Index: 14,3228

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR