4-Isopropylbenzonitrile 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A11954-03
A11954-06
A11954-14
AAA11954-03EA
43.2
USD
AAA11954-03
AAA11954-06
AAA11954-14
4-Isopropylbenzonitrile 95%
4-Isopropylbenzonitrile
MDL: MFCD00052966
Beilstein Registry No.: 1932883
Purity: 95%
CAS No.: 13816-33-6
Molecular Formula: C10H11N
Formula Weight: 145.21
Refractive Index: 1.5180
Boiling Point: 88-90°/5mm
MDL No.: MFCD00052966
Formula:
C₁₀H₁₁N Boiling Pt: 88…90 °C/5 mm Storage Temperature: Ambient |
MDL Number:
MFCD00052966 CAS Number: 13816-33-6 EINECS: 237-492-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...