4-Bromoanisole 99%
Supplier: Thermo Scientific Chemicals
A11824-36
A11824-0E
A11824-22
AAA11824-36EA
152
USD
AAA11824-36
AAA11824-0E
AAA11824-22
4-Bromoanisole 99%
4-Bromoanisole
MDL: MFCD00000097
Beilstein Registry No.: 1237590
Purity: 99%
CAS No.: 104-92-7
Molecular Formula: C7H7BrO
Formula Weight: 187.04
Flash Point: 94°(201°F)
Physical Form: Liquid
Density: 1.494
Refractive Index: 1.5640
Melting Point: 10-13°
Boiling Point: 98-99°/10mm
Beilstein Reference: 1237590
Merck Reference: 14,1427
MDL No.: MFCD00000097
Formula:
C₇H₇BrO MW: 187.04 g/mol Boiling Pt: 223 Melting Pt: 10 Density: 1.494 Flash Pt: 94 °C (201 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000097 CAS Number: 104-92-7 EINECS: 203-252-1 Merck Index: 14,1427 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...