Cholesterol 95%
Supplier: Thermo Scientific Chemicals
Synonyms:
Cholest-5-en-3-ol, Cholesterin, (3Beta)-Cholest-5-en-3-ol
Component of all biological membranes. Appears to inhibit basal channel activity in the brain
Purity: 95%
CAS No.: 57-88-5
Molecular Formula: C27H46O
Formula Weight: 386.66
Physical Form: Powder
Density: 1.067
Appearance: White
Melting Point: 147-150°
Boiling Point: 360°
Beilstein Reference: 1915888
Merck Reference: 14,2201
Optical Rotation: -36° (c=2 in dioxane)
MDL No.: MFCD00003646
Component of all biological membranes. Appears to inhibit basal channel activity in the brain
Formula:
C₂₇H₄₆O MW: 386.66 g/mol Boiling Pt: 360 Melting Pt: 147…150 °C Density: 1.067 Storage Temperature: Ambient |
MDL Number:
MFCD00003646 CAS Number: 57-88-5 EINECS: 200-353-2 Merck Index: 14,2201 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...