1-Bromopentane 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10821-0B
A10821-0I
A10821-30
AAA10821-0BEA
122
USD
AAA10821-0B
AAA10821-0I
AAA10821-30
1-Bromopentane 99%
1-Bromopentane
MDL: MFCD00000267
Beilstein Registry No.: 1730981
Purity: 99%
CAS No.: 110-53-2
Molecular Formula: CH3(CH2)4Br
Formula Weight: 151.05
Flash Point: 31°(87°F)
Refractive Index: 1.4436
Odor: Ether-like
Melting Point: -95°
Boiling Point: 127-129°
Beilstein Reference: 1730981
Merck Reference: 14,602
UN No.: 1993
MDL No.: MFCD00000267
Formula:
BrH₂C(CH₂)₃CH₃ MW: 151.05 g/mol Boiling Pt: 129 Melting Pt: -95 Density: 1.215 Flash Pt: 31 °C (87 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000267 CAS Number: 110-53-2 EINECS: 203-776-0 UN: 1993 ADR: 3 III Merck Index: 14,602 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...