Trichloroacetonitrile 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A10565-18
A10565-30
A10565-0B
AAA10565-18EA
45.3
USD
AAA10565-18
AAA10565-30
AAAA10565-0B
Trichloroacetonitrile 98%
Trichloroacetonitrile
MDL: MFCD00001842
Beilstein Registry No.: 605572
Fieser: 1,1194 5,686 6,604 7,381 12,526 13,321 21,445
Purity: 98%
CAS No.: 545-06-2
Molecular Formula: C2Cl3N
Formula Weight: 144.39
Physical Form: Clear liquid
Density: 1.44
Refractive Index: 1.4410
Appearance: Colorless
Melting Point: -44°
Boiling Point: 85-86°
Beilstein Reference: 605572
Merck Reference: 14,9628
UN No.: 3276
MDL No.: MFCD00001842
Formula:
C₂Cl₃N MW: 144.39 g/mol Boiling Pt: 85.7 Melting Pt: -44 Density: 1.44 Storage Temperature: Ambient |
MDL Number:
MFCD00001842 CAS Number: 545-06-2 EINECS: 208-885-7 UN: 3276 ADR: 6.1 III Merck Index: 14,9628 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...