Phosphorous acid 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Phosphonic acid
089407-36
089407-A3
AA89407-36EA
76
USD
AA89407-36
AA89407-A3
Phosphorous acid 97%
Phosphorous acid
MDL: MFCD00137258
Soluble in water and alcohol
Purity: 97%
CAS No.: 13598-36-2
Molecular Formula: H3PO3
Formula Weight: 82.00
Physical Form: Crystalline
Density: 1.651
Melting Point: 74°
Boiling Point: 200° dec.
Solubility: Soluble in water and alcohol
Merck Reference: 14,7346
UN No.: 2834
MDL No.: MFCD00137258
Formula:
H₃O₃P MW: 82 g/mol Boiling Pt: 200 °C (dec.) Melting Pt: 74 °C Density: 1.651 Storage Temperature: Ambient |
MDL Number:
MFCD00137258 CAS Number: 13598-36-2 EINECS: 237-066-7 UN: 2834 ADR: 8 III Merck Index: 14,7346 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...