Sodium borohydride ≥98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Sodium tetrahydridoborate, Sodium tetrahydroborate
088983-22
088983-36
AA88983-22EA
143.03
USD
AA88983-22
AA88983-36
Sodium borohydride ≥98%
Sodium borohydride
MDL: MFCD00003518
Fieser: 1,1049 15,290 16,304 17,314 18,326 19,307 20,344 21,397
Purity: 98% min
CAS No.: 16940-66-2
Molecular Formula: BH4Na
Formula Weight: 37.83
Physical Form: -4+16 Mesh Granules
Density: 1.074
Appearance: White
Odor: Weak
Melting Point: 400° dec.
Merck Reference: 14,8592
UN No.: 1426
MDL No.: MFCD00003518
Formula:
NaBH₄ Melting Pt: 400 °C (dec.) Density: 1.074 Flash Pt: 69 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003518 CAS Number: 16940-66-2 EINECS: 241-004-4 UN: 1426 ADR: 4.3,I Merck Index: 14,8592 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...