Niobium(V) chloride ≥99% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Niobium pentachloride
051108-09
051108-36
051108-22
AA51108-09EA
48.9
USD
AA51108-09
AA51108-36
AA51108-22
Niobium(V) chloride ≥99% (metals basis)
Niobium (V) chloride
In preparation of pure niobium, and as an intermediate
Purity: 99%
CAS No.: 10026-12-7
Molecular Formula: NbCl5
Formula Weight: 270.20
Physical Form: 15mm & Down Lumps, Packaged under argon
Density: 2.74
Melting Point: 208.3°
Boiling Point: 248.2°
Solubility: Decomposes in water. Soluble in alcohol, HCl, and Carbon tetrachloride
Merck Reference: 14,6557
UN No.: 3260
MDL No.: MFCD00011127
In preparation of pure niobium, and as an intermediate
Formula:
Cl₅Nb Boiling Pt: 248.2 °C Melting Pt: 208.3 °C Density: 2.74 Storage Temperature: Ambient |
MDL Number:
MFCD00011127 CAS Number: 10026-12-7 EINECS: 233-059-8 UN: 3260 ADR: 8,III Merck Index: 14,6557 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...