n-Heptane, anhydrous 99+%, chemSeal™
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
high purity solvents
Argon_Chemseal
044457-K2
044457-AE
AA44457-K2EA
84.6
USD
AA44457-K2
AA44457-AE
n-Heptane, anhydrous 99+%, chemSeal™
n-Heptane
MDL: MFCD00009544
Beilstein Registry No.: 1730763
Notes: Contains less than 100 pp H2O
Purity: 99+%
CAS No.: 142-82-5
Molecular Formula: C7H16
Formula Weight: 100.21
Flash Point: -4°(24°F)
Physical Form: Liquid
Density: 0.684
Refractive Index: 1.3877
Appearance: Colorless
Odor: Petroleum-like
Melting Point: -91°
Boiling Point: 97-99°
Beilstein Reference: 1730763
Merck Reference: 14,4659
UN No.: 1206
MDL No.: MFCD00009544
Formula:
H₃C(CH₂)₅CH₃ MW: 100.20 g/mol Boiling Pt: 98.4 Melting Pt: -90.6 Density: 0.684 Flash Pt: –4 °C (24 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009544 CAS Number: 142-82-5 EINECS: 205-563-8 UN: 1206 ADR: 3 II Merck Index: 14,4659 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...