Cobalt(II) oxide ≥99.995% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Cobalt oxide

044354-22 044354-14 044354-06
AA44354-22EA 1846 USD
AA44354-22 AA44354-14 AA44354-06
Cobalt(II) oxide ≥99.995% (metals basis)
Cobalt (II) oxide

MDL: MFCD00016031


Purity: 99.995% CAS No.: 1307-96-6 Molecular Formula: CoO, may contain up to 10% Co3O4 Formula Weight: 74.93 Physical Form: Powder Density: 6.45 Appearance: Green-brown Melting Point: 1935° Merck Reference: 14,2446 UN No.: 2811 MDL No.: MFCD00016031

Formula: CoO
MW: 74.93 g/mol
Melting Pt: 1933
Density: 6.45
Storage Temperature: Ambient
MDL Number: MFCD00016031
CAS Number: 1307-96-6
EINECS: 215-154-6
UN: 2811
ADR: 6.1 III
Merck Index: 14,2446

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR